The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000420.2 | NP_000411.1 | 12 - 13 | 1621 | 87 |
XM_005249993.2 | XP_005250050.1 | 12 - 13 | 1472 | 87 | |
XM_005249994.3 | XP_005250051.1 | 6 - 7 | 792 | 87 | |
GenBank mRNA | AK314831.1 | - | 12 - 13 | 1522 | 87 |
BC003135.1 | - | 12 - 13 | 1621 | 87 | |
BC050639.1 | - | 12 - 13 | 1516 | 87 | |
FN998902.1 | - | 11 - 12 | 1411 | 87 | |
HE572750.1 | - | 11 - 12 | 1411 | 87 | |
M64934.2 | - | 12 - 13 | 1531 | 87 |