Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KEL
Entrez Gene ID
3792
Gene Name
Kell blood group, metallo-endopeptidase
Gene Aliases
CD238, ECE3
Chromosome Location
Chr.7: 142941114 - 142962416 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000420.2 NP_000411.1 13 - 14 1706 66
XM_005249993.2 XP_005250050.1 13 - 14 1557 66
XM_005249994.3 XP_005250051.1 7 - 8 877 66
GenBank mRNA AK314831.1 - 13 - 14 1607 66
BC003135.1 - 13 - 14 1706 66
BC050639.1 - 13 - 14 1601 66
FN998902.1 - 12 - 13 1496 66
HE572750.1 - 12 - 13 1496 66
M64934.2 - 13 - 14 1616 66

View Full Product Details