Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KEL
Entrez Gene ID
3792
Gene Name
Kell blood group, metallo-endopeptidase
Gene Aliases
CD238, ECE3
Chromosome Location
Chr.7: 142941114 - 142962416 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000420.2 NP_000411.1 17 - 18 2150 95
XM_005249993.2 XP_005250050.1 17 - 18 2001 95
XM_005249994.3 XP_005250051.1 11 - 12 1321 95
GenBank mRNA AK314831.1 - 17 - 18 2051 95
BC003135.1 - 17 - 18 2150 95
BC050639.1 - 17 - 18 2045 95
FN998902.1 - 16 - 17 1940 95
HE572750.1 - 16 - 17 1940 95
KC922434.1 - 2 - 3 285 95
M64934.2 - 17 - 18 2060 95

View Full Product Details