Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CECR5
Entrez Gene ID
27440
Gene Name
cat eye syndrome chromosome region, candidate 5
Gene Aliases
-
Chromosome Location
Chr.22: 17137520 - 17165287 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017829.5 NP_060299.4 7 - 8 885 108
NM_033070.2 NP_149061.1 7 - 8 960 108
XM_005261236.3 XP_005261293.1 7 - 8 938 108
XM_011546126.2 XP_011544428.1 6 - 7 756 108
XM_011546127.1 XP_011544429.1 7 - 8 1375 108
XM_017028781.1 XP_016884270.1 7 - 8 1427 108
XM_017028782.1 XP_016884271.1 8 - 9 1754 108
XM_017028783.1 XP_016884272.1 8 - 9 2191 108
XM_017028784.1 XP_016884273.1 8 - 9 1754 108
GenBank mRNA AF273270.1 - 7 - 8 885 108
AF273271.1 - 7 - 8 945 108
AK000461.1 - 7 - 8 958 108
AK123646.1 - 2 - 3 1441 108
AK315156.1 - 7 - 8 946 108
AL833450.1 - 2 - 3 2776 108
BC042540.1 - 7 - 8 944 108

View Full Product Details