The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166111.1 | NP_001159583.1 | 25 - 26 | 3204 | 81 |
NM_001166112.1 | NP_001159584.1 | 25 - 26 | 3139 | 81 | |
NM_001166113.1 | NP_001159585.1 | 26 - 27 | 3091 | 81 | |
NM_001166114.1 | NP_001159586.1 | 23 - 24 | 3020 | 81 | |
NM_006702.4 | NP_006693.3 | 26 - 27 | 3103 | 81 | |
GenBank mRNA | AJ004832.1 | - | 26 - 27 | 2977 | 81 |
AK294021.1 | - | 25 - 26 | 3161 | 81 | |
AK302462.1 | - | 25 - 26 | 3139 | 81 | |
BC038229.1 | - | 23 - 24 | 2837 | 81 | |
BC050553.1 | - | 26 - 27 | 3066 | 81 | |
BC051768.1 | - | 25 - 26 | 3009 | 81 |