The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166111.1 | NP_001159583.1 | 4 - 5 | 583 | 88 |
NM_001166112.1 | NP_001159584.1 | 5 - 6 | 599 | 88 | |
NM_001166113.1 | NP_001159585.1 | 5 - 6 | 470 | 88 | |
NM_001166114.1 | NP_001159586.1 | 2 - 3 | 402 | 88 | |
NM_006702.4 | NP_006693.3 | 5 - 6 | 482 | 88 | |
GenBank mRNA | AJ004832.1 | - | 5 - 6 | 356 | 88 |
AK293807.1 | - | 2 - 3 | 119 | 88 | |
AK294021.1 | - | 4 - 5 | 540 | 88 | |
AK302462.1 | - | 5 - 6 | 599 | 88 | |
BC038229.1 | - | 2 - 3 | 219 | 88 | |
BC050553.1 | - | 5 - 6 | 445 | 88 | |
BC051768.1 | - | 4 - 5 | 388 | 88 | |
DA096881.1 | - | 2 - 3 | 403 | 88 | |
DB068563.1 | - | 4 - 5 | 439 | 88 |