The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319053.1 | NP_001305982.1 | 11 - 12 | 1545 | 63 |
NM_001470.3 | NP_001461.1 | 11 - 12 | 1609 | 63 | |
NM_021903.2 | NP_068703.1 | 6 - 7 | 1409 | 63 | |
NM_021904.3 | NP_068704.2 | 10 - 11 | 1423 | 63 | |
XM_005248982.2 | XP_005249039.1 | 11 - 12 | 1424 | 63 | |
XM_006715047.3 | XP_006715110.1 | 10 - 11 | 1224 | 63 | |
XM_011514453.2 | XP_011512755.1 | 9 - 10 | 1203 | 63 | |
XM_011514455.1 | XP_011512757.1 | 11 - 12 | 1424 | 63 | |
XM_017010676.1 | XP_016866165.1 | 7 - 8 | 955 | 63 | |
GenBank mRNA | AB208790.1 | - | 11 - 12 | 1344 | 63 |
AF099148.1 | - | 11 - 12 | 1431 | 63 | |
AF301005.1 | - | 10 - 11 | 1326 | 63 | |
AJ012185.1 | - | 10 - 11 | 1323 | 63 | |
AJ012186.1 | - | 6 - 7 | 972 | 63 | |
AJ012187.1 | - | 9 - 10 | 1137 | 63 | |
AJ012288.1 | - | 11 - 12 | 1366 | 63 | |
AJ225028.1 | - | 11 - 12 | 1557 | 63 | |
AJ225029.1 | - | 6 - 7 | 1151 | 63 | |
AK290191.1 | - | 6 - 7 | 1194 | 63 | |
AK312308.1 | - | 6 - 7 | 1190 | 63 | |
BC041332.2 | - | 6 - 7 | 1194 | 63 | |
BC042598.1 | - | 6 - 7 | 1183 | 63 | |
BC050532.1 | - | 6 - 7 | 1181 | 63 | |
Y11044.1 | - | 10 - 11 | 1320 | 63 |