The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319053.1 | NP_001305982.1 | 3 - 4 | 580 | 83 |
NM_001470.3 | NP_001461.1 | 3 - 4 | 580 | 83 | |
XM_005248982.2 | XP_005249039.1 | 3 - 4 | 395 | 83 | |
XM_011514455.1 | XP_011512757.1 | 3 - 4 | 395 | 83 | |
GenBank mRNA | AB208790.1 | - | 3 - 4 | 379 | 83 |
AF099148.1 | - | 3 - 4 | 402 | 83 | |
AF301005.1 | - | 2 - 3 | 297 | 83 | |
AJ012185.1 | - | 2 - 3 | 294 | 83 | |
AJ012288.1 | - | 3 - 4 | 337 | 83 | |
AJ225028.1 | - | 3 - 4 | 528 | 83 | |
Y11044.1 | - | 2 - 3 | 291 | 83 |