The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001470.3 | NP_001461.1 | 5 - 6 | 788 | 66 |
XM_005248982.2 | XP_005249039.1 | 5 - 6 | 603 | 66 | |
XM_011514455.1 | XP_011512757.1 | 5 - 6 | 603 | 66 | |
GenBank mRNA | AF099148.1 | - | 5 - 6 | 610 | 66 |
AF301005.1 | - | 4 - 5 | 505 | 66 | |
AJ012185.1 | - | 4 - 5 | 502 | 66 | |
AJ012288.1 | - | 5 - 6 | 545 | 66 | |
AJ225028.1 | - | 5 - 6 | 736 | 66 | |
Y11044.1 | - | 4 - 5 | 499 | 66 |