The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030907.3 | NP_112169.2 | 2 - 3 | 138 | |
XM_011542126.2 | XP_011540428.1 | 2 - 3 | 138 | ||
GenBank mRNA | BC002946.2 | - | 2 - 3 | 475 | 138 |
BC008702.1 | - | 2 - 3 | 475 | 138 |