Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1orf94
Entrez Gene ID
84970
Gene Name
chromosome 1 open reading frame 94
Gene Aliases
-
Chromosome Location
Chr.1: 34166883 - 34219131 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032884.4 NP_116273.2 1 - 2 290 80
GenBank mRNA AK123355.1 - 1 - 2 290 80
BC007637.1 - 1 - 2 58 80
CN366753.1 - 1 - 2 218 80
DA714248.1 - 1 - 2 150 80

View Full Product Details