Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FXYD3
Entrez Gene ID
5349
Gene Name
FXYD domain containing ion transport regulator 3
Gene Aliases
MAT8, PLML
Chromosome Location
Chr.19: 35115821 - 35124324 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136007.1 NP_001129479.1 8 - 9 626 59
NM_001136011.1 NP_001129483.1 7 - 8 504 59
NM_005971.3 NP_005962.1 6 - 7 420 59
XM_017026877.1 XP_016882366.1 6 - 7 370 59
XM_017026878.1 XP_016882367.1 5 - 6 290 59
GenBank mRNA BC005238.1 - 5 - 6 234 59
BC090044.1 - 6 - 7 369 59
BE793135.1 - 4 - 5 194 59
BI715233.1 - 5 - 6 210 59
BQ676323.1 - 7 - 8 452 59
BT006712.1 - 4 - 5 178 59
BU185141.1 - 6 - 7 307 59
CR456945.1 - 4 - 5 178 59
CR542197.1 - 4 - 5 178 59
X93036.1 - 5 - 6 237 59

View Full Product Details