Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FXYD3
Entrez Gene ID
5349
Gene Name
FXYD domain containing ion transport regulator 3
Gene Aliases
MAT8, PLML
Chromosome Location
Chr.19: 35115821 - 35124324 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136007.1 NP_001129479.1 9 - 10 660 96
NM_001136011.1 NP_001129483.1 8 - 9 538 96
NM_001136012.1 NP_001129484.1 6 - 7 532 174
NM_005971.3 NP_005962.1 7 - 8 454 96
NM_021910.2 NP_068710.1 7 - 8 616 174
XM_005258996.4 XP_005259053.1 5 - 6 437 174
XM_011527032.2 XP_011525334.1 8 - 9 745 174
XM_017026877.1 XP_016882366.1 7 - 8 404 96
XM_017026878.1 XP_016882367.1 6 - 7 324 96
GenBank mRNA BC005238.1 - 6 - 7 268 96
BC090044.1 - 7 - 8 403 96
BE793135.1 - 5 - 6 228 96
BI715233.1 - 6 - 7 244 96
BM549935.1 - 4 - 5 344 174
BQ676323.1 - 8 - 9 486 96
BT006712.1 - 5 - 6 212 96
BU185141.1 - 7 - 8 341 96
CB128589.1 - 5 - 6 345 174
CR456945.1 - 5 - 6 212 96
CR542197.1 - 5 - 6 212 96
U28249.1 - 7 - 8 543 174
X93036.1 - 6 - 7 271 96

View Full Product Details