The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293213.1 | NP_001280142.1 | 1 - 2 | 363 | 104 |
NM_178014.3 | NP_821133.1 | 1 - 2 | 363 | 104 | |
NR_120608.1 | 1 - 2 | 363 | 104 | ||
GenBank mRNA | AB062393.1 | - | 1 - 2 | 189 | 104 |
AF070561.1 | - | 1 - 2 | 184 | 104 | |
AF070593.1 | - | 1 - 2 | 184 | 104 | |
AF070600.1 | - | 1 - 2 | 201 | 104 | |
AF141349.1 | - | 1 - 2 | 201 | 104 | |
AK098772.1 | - | 1 - 2 | 208 | 104 | |
AK297489.1 | - | 1 - 2 | 117 | 104 | |
AK297638.1 | - | 1 - 2 | 212 | 104 | |
AK302195.1 | - | 1 - 2 | 212 | 104 | |
AK303227.1 | - | 1 - 2 | 223 | 104 | |
BC001938.1 | - | 1 - 2 | 188 | 104 | |
BC002347.2 | - | 1 - 2 | 193 | 104 | |
BC005838.2 | - | 1 - 2 | 188 | 104 | |
BC007605.1 | - | 1 - 2 | 193 | 104 | |
BC013374.2 | - | 1 - 2 | 184 | 104 | |
BC019924.1 | - | 1 - 2 | 181 | 104 | |
BC020946.1 | - | 1 - 2 | 222 | 104 | |
BC021909.1 | - | 1 - 2 | 183 | 104 | |
BC062532.1 | - | 1 - 2 | 181 | 104 | |
BC070326.1 | - | 1 - 2 | 199 | 104 | |
BC103746.1 | - | 1 - 2 | 206 | 104 | |
BE245819.1 | - | 1 - 2 | 217 | 104 | |
BU557299.1 | - | 1 - 2 | 363 | 104 | |
DC366174.1 | - | 1 - 2 | 346 | 104 |