The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015270.4 | NP_056085.1 | 11 - 12 | 2178 | 103 |
XM_006719210.3 | XP_006719273.1 | 11 - 12 | 3136 | 103 | |
GenBank mRNA | AB007882.2 | - | 11 - 12 | 2119 | 103 |
AF250226.1 | - | 10 - 11 | 2677 | 103 | |
BC064923.1 | - | 12 - 13 | 2100 | 103 |