The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015270.4 | NP_056085.1 | 17 - 18 | 2857 | 96 |
XM_006719210.3 | XP_006719273.1 | 17 - 18 | 3815 | 96 | |
XM_017018743.1 | XP_016874232.1 | 13 - 14 | 3019 | 96 | |
GenBank mRNA | AB007882.2 | - | 16 - 17 | 2639 | 96 |
AF250226.1 | - | 16 - 17 | 3356 | 96 | |
AK294446.1 | - | 4 - 5 | 852 | 96 | |
BC064923.1 | - | 18 - 19 | 2779 | 96 | |
BX537664.1 | - | 3 - 4 | 777 | 96 |