The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015270.4 | NP_056085.1 | 7 - 8 | 1731 | 63 |
XM_006719210.3 | XP_006719273.1 | 7 - 8 | 2689 | 63 | |
XM_017018743.1 | XP_016874232.1 | 6 - 7 | 2196 | 63 | |
GenBank mRNA | AB007882.2 | - | 7 - 8 | 1672 | 63 |
AF250226.1 | - | 6 - 7 | 2230 | 63 | |
BC064923.1 | - | 8 - 9 | 1653 | 63 |