The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032856.3 | NP_116245.2 | 1 - 2 | 106 | 83 |
NR_130945.1 | 1 - 2 | 106 | 83 | ||
NR_130947.1 | 1 - 2 | 106 | 83 | ||
GenBank mRNA | BC063392.1 | - | 1 - 2 | 74 | 83 |