Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR73
Entrez Gene ID
84942
Gene Name
WD repeat domain 73
Gene Aliases
GAMOS, HSPC264
Chromosome Location
Chr.15: 84640682 - 84654343 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032856.3 NP_116245.2 6 - 7 581 60
NR_130944.1 5 - 6 1018 60
NR_130946.1 6 - 7 588 60
NR_130947.1 5 - 6 492 60
GenBank mRNA AF161382.1 - 6 - 7 512 60
AK027794.1 - 6 - 7 518 60
AK055578.1 - 5 - 6 1018 60
AK055729.1 - 5 - 6 1100 60
AK225068.1 - 6 - 7 521 60
AK225199.1 - 6 - 7 518 60
BC014115.2 - 6 - 7 519 60
BC050648.1 - 6 - 7 538 60
BC063392.1 - 6 - 7 549 60
CD514627.1 - 5 - 6 415 60

View Full Product Details