Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACBD4
Entrez Gene ID
79777
Gene Name
acyl-CoA binding domain containing 4
Gene Aliases
HMFT0700
Chromosome Location
Chr.17: 45132600 - 45144181 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135705.2 NP_001129177.1 8 - 9 1000 95
NM_001135706.2 NP_001129178.1 8 - 9 1038 95
NM_001135707.2 NP_001129179.1 8 - 9 990 95
NM_001321352.1 NP_001308281.1 10 - 11 1162 95
NM_001321353.1 NP_001308282.1 10 - 11 1159 95
NM_024722.3 NP_078998.1 7 - 8 1154 95
XM_006722085.2 XP_006722148.1 9 - 10 1373 95
XM_017025084.1 XP_016880573.1 9 - 10 1429 95
XM_017025085.1 XP_016880574.1 10 - 11 1180 95
XM_017025086.1 XP_016880575.1 11 - 12 1329 95
XM_017025087.1 XP_016880576.1 9 - 10 1411 95
XM_017025088.1 XP_016880577.1 9 - 10 1408 95
XM_017025089.1 XP_016880578.1 9 - 10 1373 95
XM_017025090.1 XP_016880579.1 9 - 10 1391 95
XM_017025091.1 XP_016880580.1 9 - 10 1370 95
XM_017025092.1 XP_016880581.1 10 - 11 1495 95
XM_017025093.1 XP_016880582.1 9 - 10 1429 95
XM_017025094.1 XP_016880583.1 7 - 8 1152 95
GenBank mRNA AB073387.1 - 8 - 9 950 95
AK023384.1 - 7 - 8 1114 95
AK075104.1 - 8 - 9 943 95
AK296167.1 - 8 - 9 1025 95
AK316359.1 - 8 - 9 1038 95
BC029164.1 - 8 - 9 939 95
BC041143.1 - 10 - 11 1131 95
KU178779.1 - 7 - 8 694 95
KU178780.1 - 7 - 8 659 95
KU178781.1 - 7 - 8 656 95

View Full Product Details