The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278928.1 | NP_001265857.1 | 3 - 4 | 390 | 68 |
NM_017503.4 | NP_059973.4 | 3 - 4 | 390 | 68 | |
GenBank mRNA | AI808831.1 | - | 3 - 4 | 68 | |
BC014411.1 | - | 3 - 4 | 359 | 68 | |
BI118187.1 | - | 3 - 4 | 332 | 68 | |
BM789997.1 | - | 3 - 4 | 383 | 68 | |
BP244646.1 | - | 3 - 4 | 390 | 68 | |
BQ672017.1 | - | 1 - 2 | 37 | 68 | |
CR456753.1 | - | 3 - 4 | 342 | 68 | |
Z35094.1 | - | 3 - 4 | 359 | 68 |