The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008563.4 | NP_001008563.2 | 1 - 2 | 183 | 85 |
GenBank mRNA | AY074877.1 | - | 1 - 2 | 122 | 85 |