Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLXNB1
Entrez Gene ID
5364
Gene Name
plexin B1
Gene Aliases
PLEXIN-B1, PLXN5, SEP
Chromosome Location
Chr.3: 48403854 - 48430324 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130082.2 NP_001123554.1 14 - 15 3149 61
NM_002673.5 NP_002664.2 14 - 15 3316 61
XM_011533833.2 XP_011532135.1 14 - 15 3202 61
XM_011533834.1 XP_011532136.1 14 - 15 3191 61
XM_011533835.1 XP_011532137.1 14 - 15 3208 61
XM_011533836.1 XP_011532138.1 14 - 15 3124 61
XM_011533837.2 XP_011532139.1 14 - 15 3297 61
XM_017006630.1 XP_016862119.1 14 - 15 3124 61
XM_017006631.1 XP_016862120.1 13 - 14 6734 61
GenBank mRNA AB007867.1 - 14 - 15 3316 61
AJ011414.1 - 12 - 13 2463 61
AJ011415.1 - 13 - 14 2497 61
BC146793.1 - 14 - 15 3317 61
X87904.2 - 12 - 13 3046 61

View Full Product Details