The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130082.2 | NP_001123554.1 | 33 - 34 | 6042 | 94 |
NM_002673.5 | NP_002664.2 | 33 - 34 | 6209 | 94 | |
XM_011533833.2 | XP_011532135.1 | 33 - 34 | 6098 | 94 | |
XM_011533834.1 | XP_011532136.1 | 33 - 34 | 6087 | 94 | |
XM_011533835.1 | XP_011532137.1 | 33 - 34 | 6104 | 94 | |
XM_011533836.1 | XP_011532138.1 | 33 - 34 | 6020 | 94 | |
XM_011533837.2 | XP_011532139.1 | 33 - 34 | 6193 | 94 | |
XM_017006630.1 | XP_016862119.1 | 33 - 34 | 6020 | 94 | |
XM_017006631.1 | XP_016862120.1 | 32 - 33 | 9627 | 94 | |
GenBank mRNA | AB007867.1 | - | 33 - 34 | 6209 | 94 |
AJ011415.1 | - | 32 - 33 | 5390 | 94 | |
AK091832.1 | - | 10 - 11 | 1675 | 94 | |
AK092609.1 | - | 11 - 12 | 2165 | 94 | |
AK296776.1 | - | 15 - 16 | 1894 | 94 | |
BC146793.1 | - | 33 - 34 | 6210 | 94 | |
X87904.2 | - | 31 - 32 | 5939 | 94 |