The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037339.2 | NP_001032416.1 | 3 - 4 | 783 | 155 |
NM_001037340.2 | NP_001032417.1 | 8 - 9 | 1263 | 155 | |
NM_001037341.1 | NP_001032418.1 | 10 - 11 | 1208 | 155 | |
NM_001297440.1 | NP_001284369.1 | 9 - 10 | 969 | 155 | |
NM_001297441.1 | NP_001284370.1 | 9 - 10 | 862 | 155 | |
NM_001297442.1 | NP_001284371.1 | 3 - 4 | 787 | 155 | |
NM_002600.3 | NP_002591.2 | 10 - 11 | 1163 | 155 | |
XM_005270924.3 | XP_005270981.1 | 4 - 5 | 454 | 155 | |
XM_005270925.2 | XP_005270982.1 | 4 - 5 | 495 | 155 | |
XM_006710680.3 | XP_006710743.1 | 4 - 5 | 520 | 155 | |
XM_011541566.1 | XP_011539868.1 | 8 - 9 | 1026 | 155 | |
XM_017001445.1 | XP_016856934.1 | 7 - 8 | 615 | 155 | |
XM_017001446.1 | XP_016856935.1 | 7 - 8 | 1213 | 155 | |
GenBank mRNA | AB209081.1 | - | 8 - 9 | 1172 | 155 |
AK289969.1 | - | 10 - 11 | 1163 | 155 | |
AK290006.1 | - | 8 - 9 | 1144 | 155 | |
AK290206.1 | - | 3 - 4 | 778 | 155 | |
AK294441.1 | - | 3 - 4 | 478 | 155 | |
BC101480.1 | - | 10 - 11 | 1103 | 155 | |
BC105040.1 | - | 10 - 11 | 1103 | 155 | |
CR749667.1 | - | 9 - 10 | 968 | 155 | |
EF595686.1 | - | 3 - 4 | 318 | 155 | |
KJ534916.1 | - | 9 - 10 | 1017 | 155 | |
KJ534917.1 | - | 8 - 9 | 792 | 155 | |
KJ535082.1 | - | 8 - 9 | 778 | 155 | |
L12686.1 | - | 3 - 4 | 695 | 155 | |
L20966.1 | - | 10 - 11 | 1155 | 155 | |
M97515.1 | - | 3 - 4 | 782 | 155 | |
U85048.1 | - | 8 - 9 | 972 | 155 |