The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130156.2 | NP_001123628.1 | 4 - 5 | 585 | 93 |
NM_001130157.2 | NP_001123629.1 | 5 - 6 | 658 | 93 | |
NM_001195432.1 | NP_001182361.1 | 6 - 7 | 860 | 93 | |
NM_001195434.1 | NP_001182363.1 | 6 - 7 | 703 | 93 | |
NM_022443.4 | NP_071888.1 | 4 - 5 | 541 | 93 | |
XM_005247479.4 | XP_005247536.1 | 5 - 6 | 588 | 93 | |
XM_005247480.2 | XP_005247537.1 | 5 - 6 | 572 | 93 | |
XM_006713645.2 | XP_006713708.1 | 3 - 4 | 379 | 93 | |
XM_011512852.2 | XP_011511154.1 | 6 - 7 | 661 | 93 | |
XM_011512853.2 | XP_011511155.1 | 5 - 6 | 608 | 93 | |
XM_011512854.2 | XP_011511156.1 | 3 - 4 | 377 | 93 | |
GenBank mRNA | AK056948.1 | - | 6 - 7 | 673 | 93 |
AK096889.1 | - | 6 - 7 | 769 | 93 | |
AK297488.1 | - | 4 - 5 | 527 | 93 | |
AK315402.1 | - | 4 - 5 | 511 | 93 | |
AY848702.1 | - | 4 - 5 | 554 | 93 | |
BC007045.1 | - | 4 - 5 | 510 | 93 | |
BX647666.1 | - | 5 - 6 | 596 | 93 | |
KU178186.1 | - | 4 - 5 | 404 | 93 | |
KU178187.1 | - | 5 - 6 | 449 | 93 | |
KU178188.1 | - | 3 - 4 | 256 | 93 | |
L49054.1 | - | 4 - 5 | 512 | 93 |