Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRN
Entrez Gene ID
2896
Gene Name
granulin
Gene Aliases
CLN11, GEP, GP88, PCDGF, PEPI, PGRN
Chromosome Location
Chr.17: 44345086 - 44353106 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002087.3 NP_002078.1 3 - 4 525 73
XM_005257253.1 XP_005257310.1 3 - 4 570 73
GenBank mRNA AF055008.1 - 3 - 4 331 73
AK000607.1 - 3 - 4 309 73
AK222522.1 - 3 - 4 309 73
AK296090.1 - 3 - 4 263 73
AK303830.1 - 3 - 4 306 73
AY124489.1 - 2 - 3 287 73
BC000324.2 - 3 - 4 488 73
BC010577.2 - 2 - 3 279 73
BT006844.1 - 2 - 3 269 73
M75161.1 - 2 - 3 281 73
X62320.1 - 3 - 4 309 73

View Full Product Details