The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002087.3 | NP_002078.1 | 12 - 13 | 1902 | 75 |
XM_005257253.1 | XP_005257310.1 | 12 - 13 | 1947 | 75 | |
GenBank mRNA | AF055008.1 | - | 12 - 13 | 1708 | 75 |
AK000607.1 | - | 12 - 13 | 1686 | 75 | |
AK023348.1 | - | 9 - 10 | 1190 | 75 | |
AK222522.1 | - | 12 - 13 | 1686 | 75 | |
AK296090.1 | - | 12 - 13 | 1640 | 75 | |
AK303830.1 | - | 9 - 10 | 1212 | 75 | |
AY124489.1 | - | 11 - 12 | 1664 | 75 | |
BC000324.2 | - | 11 - 12 | 1400 | 75 | |
BC010577.2 | - | 11 - 12 | 1656 | 75 | |
BT006844.1 | - | 10 - 11 | 1181 | 75 | |
X62320.1 | - | 12 - 13 | 1686 | 75 |