The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144961.1 | NP_001138433.1 | 3 - 4 | 629 | 63 |
NM_001144963.1 | NP_001138435.1 | 3 - 4 | 576 | 63 | |
GenBank mRNA | BC143671.1 | - | 3 - 4 | 567 | 63 |