The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014972.2 | NP_001014972.1 | 5 - 6 | 2024 | 87 |
NM_001252612.1 | NP_001239541.1 | 5 - 6 | 2350 | 87 | |
NM_001252613.1 | NP_001239542.1 | 5 - 6 | 2034 | 87 | |
NM_014497.4 | NP_055312.2 | 5 - 6 | 2034 | 87 | |
XM_005264263.1 | XP_005264320.1 | 4 - 5 | 498 | 87 | |
XM_006711989.1 | XP_006712052.1 | 4 - 5 | 510 | 87 | |
XM_011532768.2 | XP_011531070.2 | 5 - 6 | 2120 | 87 | |
XM_017003809.1 | XP_016859298.1 | 5 - 6 | 2125 | 87 | |
XM_017003810.1 | XP_016859299.1 | 5 - 6 | 2035 | 87 | |
XM_017003811.1 | XP_016859300.1 | 5 - 6 | 2120 | 87 | |
XM_017003812.1 | XP_016859301.1 | 5 - 6 | 2120 | 87 | |
GenBank mRNA | AF534078.1 | - | 4 - 5 | 1714 | 87 |
AK291198.1 | - | 5 - 6 | 1998 | 87 | |
AK303723.1 | - | 3 - 4 | 494 | 87 | |
AK309221.1 | - | 5 - 6 | 2075 | 87 | |
BC024000.1 | - | 5 - 6 | 1993 | 87 | |
BC083513.1 | - | 5 - 6 | 1993 | 87 | |
BC143728.1 | - | 5 - 6 | 1987 | 87 | |
BX537425.1 | - | 5 - 6 | 1998 | 87 | |
CR749322.1 | - | 5 - 6 | 2347 | 87 | |
D83032.1 | - | 5 - 6 | 2026 | 87 |