The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021619.2 | NP_067632.2 | 1 - 2 | 281 | 48 |
GenBank mRNA | AY004252.2 | - | 1 - 2 | 281 | 48 |