Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRDM12
Entrez Gene ID
59335
Gene Name
PR domain 12
Gene Aliases
HSAN8, PFM9
Chromosome Location
Chr.9: 130664594 - 130682997 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021619.2 NP_067632.2 2 - 3 479 76
GenBank mRNA AY004252.2 - 2 - 3 479 76

View Full Product Details