The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021619.2 | NP_067632.2 | 3 - 4 | 635 | 67 |
GenBank mRNA | AY004252.2 | - | 3 - 4 | 635 | 67 |