Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HINFP
Entrez Gene ID
25988
Gene Name
histone H4 transcription factor
Gene Aliases
HiNF-P, MIZF, ZNF743
Chromosome Location
Chr.11: 119121523 - 119135055 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243259.1 NP_001230188.1 1 - 2 117 96
NM_198971.2 NP_945322.1 1 - 2 117 96
XM_017017500.1 XP_016872989.1 1 - 2 53 96
XM_017017504.1 XP_016872993.1 1 - 2 47 96
XM_017017508.1 XP_016872997.1 1 - 2 47 96
GenBank mRNA AK300288.1 - 1 - 2 48 96
BF726036.1 - 1 - 2 62 96

View Full Product Details