Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MON1A
Entrez Gene ID
84315
Gene Name
MON1 homolog A, secretory trafficking associated
Gene Aliases
SAND1
Chromosome Location
Chr.3: 49908869 - 49930012 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032355.3 NP_115731.2 2 - 3 544 65
XM_006713345.3 XP_006713408.1 2 - 3 310 65
XM_011534160.1 XP_011532462.1 2 - 3 173 65
GenBank mRNA AK074404.1 - 2 - 3 354 65
AK315630.1 - 2 - 3 398 65
BC006299.2 - 2 - 3 356 65
BC009459.1 - 2 - 3 313 65
BQ946394.1 - 2 - 3 676 65

View Full Product Details