Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MON1A
Entrez Gene ID
84315
Gene Name
MON1 homolog A, secretory trafficking associated
Gene Aliases
SAND1
Chromosome Location
Chr.3: 49908869 - 49930012 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142501.1 NP_001135973.1 3 - 4 1312 95
NM_032355.3 NP_115731.2 4 - 5 1798 95
XM_006713345.3 XP_006713408.1 4 - 5 1564 95
XM_011534160.1 XP_011532462.1 4 - 5 1427 95
GenBank mRNA AK074404.1 - 4 - 5 1608 95
AK315630.1 - 4 - 5 1652 95
BC006299.2 - 4 - 5 1610 95
BC009459.1 - 4 - 5 1567 95
BC047022.1 - 3 - 4 1062 95

View Full Product Details