Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB43
Entrez Gene ID
23099
Gene Name
zinc finger and BTB domain containing 43
Gene Aliases
ZBTB22B, ZNF-X, ZNF297B
Chromosome Location
Chr.9: 126804072 - 126838210 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014007.3 NP_054726.1 1 - 2 123 145
GenBank mRNA AF049907.1 - 1 - 2 103 145
AK026123.1 - 1 - 2 69 145
DA864775.1 - 1 - 2 125 145

View Full Product Details