Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- GPATCH8
- Entrez Gene ID
-
23131
- Gene Name
- G-patch domain containing 8
- Gene Aliases
-
GPATC8, KIAA0553
- Chromosome Location
-
Chr.17: 44395277 - 44503602 on Build GRCh38
View Full Product Details