The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000691.4 | NP_000682.3 | 6 - 7 | 988 | 71 |
NM_001135167.1 | NP_001128639.1 | 6 - 7 | 983 | 71 | |
NM_001135168.1 | NP_001128640.1 | 5 - 6 | 1138 | 71 | |
XM_005256522.1 | XP_005256579.1 | 5 - 6 | 1546 | 71 | |
XM_005256523.2 | XP_005256580.1 | 5 - 6 | 2054 | 71 | |
XM_005256524.3 | XP_005256581.1 | 4 - 5 | 1353 | 71 | |
XM_011523730.1 | XP_011522032.1 | 5 - 6 | 752 | 71 | |
XM_011523731.1 | XP_011522033.1 | 5 - 6 | 1484 | 71 | |
GenBank mRNA | AK091272.1 | - | 3 - 4 | 2383 | 71 |
AK093755.1 | - | 5 - 6 | 1845 | 71 | |
AK093877.1 | - | 4 - 5 | 1287 | 71 | |
AK225513.1 | - | 6 - 7 | 858 | 71 | |
AK292193.1 | - | 5 - 6 | 1138 | 71 | |
AK314584.1 | - | 6 - 7 | 848 | 71 | |
BC004102.2 | - | 6 - 7 | 987 | 71 | |
BC004370.1 | - | 6 - 7 | 840 | 71 | |
BC008892.2 | - | 6 - 7 | 831 | 71 | |
BC021194.2 | - | 6 - 7 | 829 | 71 | |
BT007102.1 | - | 5 - 6 | 808 | 71 | |
M74542.1 | - | 6 - 7 | 850 | 71 | |
M77477.1 | - | 5 - 6 | 819 | 71 | |
S61044.1 | - | 5 - 6 | 808 | 71 |