The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271.3 | NP_001262.3 | 16 - 17 | 2573 | 75 |
GenBank mRNA | AF006514.1 | - | 16 - 17 | 2705 | 75 |
AK292850.1 | - | 16 - 17 | 2570 | 75 | |
DB121252.1 | - | 2 - 3 | 240 | 75 |