Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHD2
Entrez Gene ID
1106
Gene Name
chromodomain helicase DNA binding protein 2
Gene Aliases
EEOC
Chromosome Location
Chr.15: 92900321 - 93028007 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271.3 NP_001262.3 36 - 37 5270 92
GenBank mRNA AF006514.1 - 36 - 37 5402 92

View Full Product Details