The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016474.4 | NP_057558.3 | 5 - 6 | 575 | 66 |
NR_135523.1 | 5 - 6 | 575 | 66 | ||
XM_017006554.1 | XP_016862043.1 | 5 - 6 | 638 | 66 | |
XM_017006555.1 | XP_016862044.1 | 5 - 6 | 636 | 66 | |
GenBank mRNA | AK309525.1 | - | 5 - 6 | 548 | 66 |
BC013999.2 | - | 5 - 6 | 551 | 66 | |
BC033897.1 | - | 5 - 6 | 66 |