Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTDSPL
Entrez Gene ID
10217
Gene Name
CTD small phosphatase like
Gene Aliases
C3orf8, HYA22, PSR1, RBSP3, SCP3
Chromosome Location
Chr.3: 37862152 - 37984469 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017005519.1 XP_016861008.1 8 - 9 874 62
XM_017005520.1 XP_016861009.1 8 - 9 2192 62

View Full Product Details