Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF40
Entrez Gene ID
9810
Gene Name
ring finger protein 40
Gene Aliases
BRE1B, RBP95, STARING
Chromosome Location
Chr.16: 30761612 - 30800231 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207033.1 NP_001193962.1 8 - 9 1820 64
NM_001286572.2 NP_001273501.1 8 - 9 1181 64
NM_014771.3 NP_055586.1 8 - 9 1820 64
XM_011545997.1 XP_011544299.1 8 - 9 1161 64
GenBank mRNA AB014561.1 - 8 - 9 1092 64
AF122819.1 - 8 - 9 1096 64
AK075526.1 - 6 - 7 841 64
AK291666.1 - 8 - 9 1110 64
BC006133.1 - 8 - 9 1087 64
BC011769.2 - 8 - 9 1087 64
BC018647.2 - 8 - 9 1087 64
BX640763.1 - 5 - 6 454 64
CR627431.1 - 8 - 9 1149 64
KU178492.1 - 7 - 8 1000 64
KU178493.1 - 7 - 8 1000 64

View Full Product Details