Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRRT1
Entrez Gene ID
80863
Gene Name
proline rich transmembrane protein 1
Gene Aliases
C6orf31, DSPD1, IFITMD7, NG5
Chromosome Location
Chr.6: 32148363 - 32151943 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030651.3 NP_085154.3 3 - 4 863 81
GenBank mRNA AK054885.1 - 3 - 4 863 81
AK123424.1 - 2 - 3 1745 81
BC013201.1 - 5 - 6 689 81
BC063046.1 - 3 - 4 811 81

View Full Product Details