Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIRT6
Entrez Gene ID
51548
Gene Name
sirtuin 6
Gene Aliases
SIR2L6
Chromosome Location
Chr.19: 4174109 - 4182604 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193285.2 NP_001180214.1 4 - 5 501 100
NM_001321058.1 NP_001307987.1 3 - 4 373 100
NM_001321060.1 NP_001307989.1 4 - 5 501 100
NM_001321061.1 NP_001307990.1 3 - 4 373 100
NM_001321064.1 NP_001307993.1 4 - 5 466 100
NM_016539.3 NP_057623.2 4 - 5 501 100
GenBank mRNA AF233396.1 - 4 - 5 496 100
AK074810.1 - 4 - 5 464 100
AK225567.1 - 4 - 5 469 100
AK293348.1 - 3 - 4 334 100
AK299420.1 - 3 - 4 316 100
AK315048.1 - 4 - 5 465 100
AL559383.3 - 4 - 5 439 100
BC004218.2 - 3 - 4 313 100
BC005026.1 - 4 - 5 439 100
BC028220.1 - 4 - 5 447 100
CN364175.1 - 4 - 5 444 100
CN364177.1 - 4 - 5 504 100
CR457200.1 - 4 - 5 436 100

View Full Product Details