The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002811.4 | NP_002802.2 | 1 - 2 | 213 | 62 |
GenBank mRNA | AK315381.1 | - | 1 - 2 | 171 | 62 |
BC000338.2 | - | 1 - 2 | 200 | 62 | |
BC012606.1 | - | 1 - 2 | 194 | 62 | |
BU601564.1 | - | 1 - 2 | 216 | 62 |