The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080437.1 | NP_001073906.1 | 29 - 30 | 4122 | 95 |
XM_011510931.2 | XP_011509233.1 | 30 - 31 | 4436 | 95 | |
XM_011510932.2 | XP_011509234.1 | 29 - 30 | 4339 | 95 | |
XM_017003773.1 | XP_016859262.1 | 30 - 31 | 4438 | 95 | |
XM_017003774.1 | XP_016859263.1 | 29 - 30 | 4207 | 95 | |
XM_017003775.1 | XP_016859264.1 | 29 - 30 | 4341 | 95 | |
XM_017003776.1 | XP_016859265.1 | 29 - 30 | 4324 | 95 | |
XM_017003777.1 | XP_016859266.1 | 28 - 29 | 4215 | 95 | |
XM_017003778.1 | XP_016859267.1 | 29 - 30 | 4202 | 95 | |
XM_017003779.1 | XP_016859268.1 | 30 - 31 | 4138 | 95 | |
XM_017003780.1 | XP_016859269.1 | 29 - 30 | 3994 | 95 | |
XM_017003781.1 | XP_016859270.1 | 29 - 30 | 3984 | 95 | |
XM_017003782.1 | XP_016859271.1 | 25 - 26 | 3333 | 95 | |
GenBank mRNA | AF439717.1 | - | 23 - 24 | 2965 | 95 |
AK074062.1 | - | 28 - 29 | 3730 | 95 | |
BC027939.1 | - | 11 - 12 | 1861 | 95 |