The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014597.4 | NP_055412.2 | 2 - 3 | 1728 | 125 |
GenBank mRNA | AB046574.1 | - | 2 - 3 | 1708 | 125 |
AK123702.1 | - | 2 - 3 | 2968 | 125 | |
AK222722.1 | - | 2 - 3 | 1701 | 125 | |
AK292379.1 | - | 2 - 3 | 1701 | 125 | |
AY336729.1 | - | 2 - 3 | 1709 | 125 | |
AY394925.1 | - | 2 - 3 | 1694 | 125 | |
BC130622.1 | - | 2 - 3 | 1706 | 125 | |
U15552.1 | - | 2 - 3 | 1694 | 125 |