Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNTTIP2
Entrez Gene ID
30836
Gene Name
deoxynucleotidyltransferase terminal interacting protein 2
Gene Aliases
ERBP, FCF2, HSU15552, LPTS-RP2, TdIF2
Chromosome Location
Chr.1: 93869458 - 93879206 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014597.4 NP_055412.2 4 - 5 1959 115
GenBank mRNA AB046574.1 - 4 - 5 1939 115
AK222722.1 - 4 - 5 1932 115
AK292379.1 - 4 - 5 1932 115
AK302909.1 - 3 - 4 1776 115
AY336729.1 - 4 - 5 1940 115
AY394925.1 - 4 - 5 1925 115
BC130622.1 - 4 - 5 1937 115
BX648593.1 - 3 - 4 442 115
U15552.1 - 4 - 5 1925 115

View Full Product Details